Critical Filter Asbestos Vacuum, Model X-829, Minuteman®
Supplier: Minuteman
C82915-06
62510-086EA 2766.64 USD
62510-086
Critical Filter Asbestos Vacuum, Model X-829, Minuteman®
Vacuum Cleaners
Use vacuum anywhere wet or dry waste removal is needed
Specifically designed for the recovery of asbestos and other hazardous material. Features two 20.3 cm (8") dia. wheels and two 7.6 cm (3") dia. casters. The 57 L (15 gal.) rugged polyethylene tank features a self-sealing intake to ensure a positive vacuum seal while allowing hose to swivel. “Sure-grip/quick release” mechanism ensures easy connection and removal of hoses. Vacuum is equipped with a HEPA filter with an efficiency rating of 99.99% for 0.12µ. A safety interlock system with unique microswitch does not allow vacuum to start until the filter is properly aligned, protecting the motor and user from contamination. A unitized polyethylene UniAdapter™ attaches to the vacuum for conversion to leakproof, wet applications. Acoustically-designed polypropylene lid minimizes noise. Conforms to all U.S. EPA and OSHA guidelines and requirements for cleaning hazardous materials and filtering all toxic and noxious dust. (Minuteman C82915-06)
Ordering information: Supplied with tank and UniAdapter.