SP Bel-Art Stainless Steel Reagent Digger, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F36713-0000
62489-110CS 284.78 USD
62489-110
SP Bel-Art Stainless Steel Reagent Digger, Bel-Art Products, a part of SP
Scoops
Ideal for dislodging and retrieving stubborn, caked, or crystallized materials from deep down inside a bottle or vessel that has a minimum neck diameter of 24 mm (15/16").
  • Ergonomically designed
  • Suitable for a variety of reagent applications
  • Stainless steel construction
  • Ideal for removing stubborn materials from deep vessels
This utensil is ergonomically designed with a curved shape, one pointed end, and one blunt end. The shape facilitates scooping, shoveling, digging, and transferring of powdered, flaked, crystalline, or granular media.