Nunc-Immuno™ TSP Replicator, MaxiSorp™, Thermo Scientific
Supplier: Thermo Fisher Scientific
445497
62409-179CS 632.17 USD
62409-179
Nunc-Immuno™ TSP Replicator, MaxiSorp™, Thermo Scientific
Microplate and Petri Dish Replicators
This Transferable Solid Phase (TSP) disposable replicator is similar to a 96-well plate lid with 96 attached pins.
  • Suitable for the adsorption of mixed proteins or antibodies directly from cultures
  • Sterilized, polystyrene
When precoated with antigen or antibodies, a specific analyte from a culture may be assayed, via efficient transfer from one plate to another. Can be assayed in the Nunc MicroWell™ 96-well plates.
When placed over the well plate, the pins are immersed in the wells. MaxiSorp™ is ideal for replating and screening of 96-well plates containing transformants, library clones, screening hybridoma cultures, and expression libraries.