Nunc® Polyester Sealing Tape, Thermo Scientific
Supplier: Thermo Fisher Scientific
236366
62409-124CS 1292.42 USD
62409-124
Nunc® Polyester Sealing Tape, Thermo Scientific
Seals Microplate Seals
Polyester film with acrylic adhesive.
  • Minimum evaporation and well-to-well cross-contamination
  • Non-pierceable
  • Applicator dimensions: 80 x 134 mm
Provides an effective seal for all MicroWell™ plates used in PCR and cell culture applications.