Nalgene® Stainless Steel Storage Racks for Microplates, Thermo Scientific
Supplier: Thermo Fisher Scientific
5039-0048
62409-082CS 644.08 USD
62409-082
Nalgene® Stainless Steel Storage Racks for Microplates, Thermo Scientific
Racks Microplate Racks
Nalgene® Stainless Steel Storage racks for Micro plates come in two convenient horizontal sizes to fit your freezer or refrigerator
These storage racks have sturdy handles on top and on the sides for easy transport. Each compartment on the racks comes with a retainer clip which holds the plates securely in place.