Corning® BioCoat™ Intestinal Epithelium Differentiation Environment, Corning
Supplier: Corning
355057
62405-098EA 580.35 USD
62405-098
Corning® BioCoat™ Intestinal Epithelium Differentiation Environment, Corning
Cell Separation Media
Corning BioCoat™ Intestinal Epithelium Differentiation Environment is an integrated system designed to create in vitro intestinal models.
Includes 24 Corning® BioCoat™ Fibrillar Collagen Cell Culture Inserts (1µm pore size in two 24-well plates), 100mL Intestinal Epithelium Seeding Basal Medium, 400mL Intestinal Epithelium Differentiation Medium, and 0.5mL MITO+ Serum Extender.