Microlite™ Microtiter® 96-Well Plates and Strips, Thermo Scientific
Supplier: Thermo Fisher Scientific
7417-12
62402-974CS 432.61 USD
62402-974
Microlite™ Microtiter® 96-Well Plates and Strips, Thermo Scientific
Microplates Immunology Microplates and Strips
These flat-bottom plates were developed for luminescence applications, including PCR quantification, report gene detection, phagocytosis and luminescent immunoassay.
Made of a densely pigmented, white polystyrene formulation developed for luminescent assays, Microlite™ 1 is ideal for applications requiring high reflectivity and minimal crosstalk. Microlite™ 2, irradiated, is ideal for applications requiring higher binding with the same high reflectivity and minimal crosstalk.