VWR® Light Meter with Outputs
Supplier: VWR International
90027-68
62344-944EA 286.89 USD
62344-944
VWR® Light Meter with Outputs
Light Meters
Dual range light meter with outputs reads both Lux and Foot Candles on a large 1.27 cm (1/2") LCD.
  • Traceable®
  • Dual range
  • Analog and digital output
  • Fast response
Analog output of 1mV per digit allows readings to be captured by any chart recorder, strip recorder, or computer which accepts mV signals. Features fast response sampling rate of 0.4 seconds, automatic zero adjustment, selectable display update rate of 1 or 2 seconds, and data hold switch to freeze readings.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Unit is supplied with photo cell, dual recorder outputs, and 9V battery.