VWR® Redi-Stor™ Conductivity Probe Storage Solution
Supplier: VWR International
62344-940
62344-940EA 84.29 USD
62344-940
VWR® Redi-Stor™ Conductivity Probe Storage Solution
Electrolytes Electrode Storage Solutions
Ideal solution for storing conductivity probes.
  • No conditioning required after removal from storage
  • Keeps probe clean
Probe stays clean and ready for immediate use with no conditioning necessary. Eliminates growths common when storing in only water.