VWR® Traceable® Thermometer/Clock/Humidity Monitor
Supplier: VWR International
62344-734 10754-720 10754-740
62344-734EA 118.72 USD
62344-734 10754-720 10754-740
VWR® Traceable® Thermometer/Clock/Humidity Monitor
Thermometers Digital Thermometers
Unit simultaneously displays ambient temperature, humidity, and time of day.
  • Traceable®
  • High/low temperature and humidity memory
  • Easy-to-read display
Ideal for use in incubators, storerooms, drying chambers, hoods, cleanrooms, and environmental cabinets.
Memory recalls highest and lowest temperature and humidity readings. Clock displays a.m./p.m. or 24-hour military time to the exact minute with 0.01% accuracy.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with one-year AAA battery, built-in stand, and wall mount.