VWR® Mini-Alarm Timer/Stopwatch
Supplier: VWR International
90013-07
62344-585EA 81.06 USD
62344-585
VWR® Mini-Alarm Timer/Stopwatch
Timers Digital Stopwatches
Use individually numbered keys to quickly set countdown time to any value up to 99 minutes, 99 seconds.
  • Traceable®
  • LCD digits are readable from 2.4 m (8') away
  • Rugged, chemical-resistant ABS plastic case
Stopwatch mode with time-out feature counts up to 59 minutes, 59 seconds, then automatically rolls over to zero to continue timing.
Unit incorporates a flip-open easel for benchtop use, a spring fastener for clipping to lab coat, and a magnetic back for use with incubators, refrigerators, ovens, and more.
High decibel countdown alarm turns off manually or automatically after one minute.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a two-year, 1.5 V silver-oxide battery.