Model VT60 Series Wet/Dry HEPA Vacuum with Accessory Kit, Nilfisk
Supplier: Nilfisk
01799531
62111-348EA 9553.65 USD
62111-348 76249-122
Model VT60 Series Wet/Dry HEPA Vacuum with Accessory Kit, Nilfisk
Vacuum Cleaners
The Nilfisk VT60 wet/dry vacuum simultaneously picks up wet and dry debris without special adapters or filter switching.
  • Multi-stage filtration system designed for liquid or dry pick-up, with no filter change or adjustment
  • Detachable trolley features sturdy polyethylene construction and large, smooth wheels for easy decontamination and added maneuverability
  • Foam impact filter fits over and protects the float valve from clogging debris
  • Pliable rubber inlet tube designed to relieve caking and clogging caused by collection of damp or sticky material
Featuring a unique four-stage filtration system and multiple collection and disposal options, the versatility of the VT60 is ideal for multiple wet/dry applications.
Model VT 60CR is designed specifically for cleanroom applications.
Ordering information: Accessories included are plastic hose assembly, aluminum wand (qty. 2), dust brush, universal nozzle-3 inserts, crevice nozzle, package of (25) polyliners.
Packaging: Accessories could add additional weight and boxes to the order.