Ground Cord, SCS, Desco Industries
Supplier: 3M
054007-34649 3048
62111-218EA 21.13 USD
62111-218 62111-223
Ground Cord, SCS, Desco Industries
Grounders Grounding Wires
Ground Cords easily snap onto mats and runners.
  • 15' (4.6 m) ground cord with 10 mm snap
  • Connects all static control mats and runners to building ground
Ground cord has a one megohm resistor for mat connection only.