Single Wire Disposable Wrist Strap, SCS, Desco Industries
Supplier: 3M
051138-37890
62111-190EA 2.33 USD
62111-190
Single Wire Disposable Wrist Strap, SCS, Desco Industries
Grounders Wrist Grounders
2209 Wrist Strap is designed for one-time use. The wrist strap is ideally suited for shipment along with ESD sensitive items [ESDS], providing the means to ground personnel opening ESDS packaging and installing contents.
  • Disposable non-woven fabric wrist strap provides cost-effective
  • Hypo-allergenic adhesive provides 360 degree contact with skin
  • Minimizes skin-to-band resistance
  • Conductive adhesive on the copper foil adheres to any convenient electrical ground
Use for visitors and one-time use applications. Suited for shipment along with components and devices, such as memory upgrades, sound cards, etc., to provide protection at the receiving end. Equipment chassis to provide equipotential bonding. A current limiting resistance is fabricated into the plastic ribbon below the wristband.
Certifications: Meets the ANSI/ESD S20.20 wrist strap system required limit tested per ESD TR53. UL listed.
Packaging: Individually packaged.