SP Bel-Art H-B DURAC® Blood Bank Liquid-In-Glass Refrigerator Thermometer, Organic Liquid Fill, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
B60805-0100
61222-617EA 100.23 USD
61222-617
SP Bel-Art H-B DURAC® Blood Bank Liquid-In-Glass Refrigerator Thermometer, Organic Liquid Fill, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
This total immersion thermometer indicates critical temperature ranges for blood bank refrigerators and vaccine storage
Critical temperatures (4°C and 6°C) are marked with arrows. The thermometer contains red organic liquid against white back glass, and features PFA flouropolymer safety coating to keep liquid and glass encapsulated in the event of breakage.