VWR® Traceable® Digital Thermometer

Supplier: VWR International
90008-06 VC4000 VC1009 VC2009
61220-601EA 1459.17 USD
61220-601 10171-226 10171-390 10171-530
VWR® Traceable® Digital Thermometer
Thermometers Digital Thermometers
Ideal for measuring temperatures in cuvettes, gas systems, reagents, and chemical solutions, or for routine measurements.

  • Traceable®
  • Read °C and °F
  • Compact and portable
  • Resolution of 0.001°

High-contrast LCD features 12.7mm (1/2) digits

Units Read °C or °F at the touch of a button.

The Data Acquisition System captures, displays, and stores readings in Windows®. Data can be read as-is or imported into databases, spreadsheets, word processors, and statistical programs.

Readings are updated three times a second. Features include hold button, minimum/maximum memories, and 50 data logger memories. Units operate on 9V alkaline battery (included) or optional 115V adapter/recharger.Thermometer 61220-601 comes with a standard quick-response, stainless steel triple purpose probe that features a 1.5m (5') cable and can be used with liquids, semi-solids, or air/gas.

Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

Accessories information: The Traceable® thermistor accessories are designed to be used with thermistor thermometer 61220-601.

Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.

For improved performance, probes should be calibrated as a system with the meter.

Ordering information: Thermometer 61220-601 is supplied with probe and 1.5m (5') cable, 9V battery, carrying case, and manual. The Data Acquisition System is supplied with an AC adapter, 8.9 cm (31/2) diskette, and 91.4 cm (36") cable that plugs into the instrument and computer serial port.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR