Safety Knife, SECUNORM HANDY, MARTOR
Supplier: Martor
1444.02
61200-338EA 24.15 USD
61200-338
Safety Knife, SECUNORM HANDY, MARTOR
Knives
The handy right and left hander knife is a semi automatic retractable spring loaded knife for cutting general industrial materials.
  • Ergonomically friendly ambidextrous slider mechanism to engage the blade
  • Cutting depth: 8.5 mm
  • Easy, tool free blade change
Safer alternative to fixed blade utility knives for general cutting tasks. This kinfe provides high safety, and very abrasion resistant.