Safety Cutters, ZEPHER, Martor
Supplier: Martor
06.08
61200-334EA 100.83 USD
61200-334
Safety Cutters, ZEPHER, Martor
Knives
Heavy-duty glass filled safety cutter for removing multiple layers of film.
  • Improved safety feature eliminates accidental cuts
This design allows the use of two hands, which provides more push power.