VWR® Handheld Digital Barometer
Supplier: VWR International
61161-398 10754-856
61161-398EA 522.38 USD
61161-398 10754-856
VWR® Handheld Digital Barometer
Barometers
Compact and lightweight handheld barometer measures barometric pressure, temperature, and altitude. It graphically displays the barometric pressure trend and features a stopwatch/time-of-day clock.
  • Stopwatch times events from 1 second to 99 hours, 59 minutes, 59 seconds
  • 12- or 24-hour clock with alarm displays the time of day to the minute and shows the date and month
  • Temperature display is updated every 10 seconds
  • Min./max. memory for the altitudes reached during any time period; display updates altitude every 2 seconds
  • Altitude trend shows present reading and previous 2, 4, 6, 8, 10, and 12 hours in bar chart format
Barometric pressure is displayed in mbar or inches. Altitude is displayed in meters or feet.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Delivery information: Supplied complete with rugged shock-resistant case, lanyard, 1.5V silver oxide battery, and instructions.