VWR® One-Piece Humidity/Temperature Thermometer
Supplier: VWR International
90029-51 VC4187
61161-380EA 528.01 USD
61161-380 10754-546
VWR® One-Piece Humidity/Temperature Thermometer
Hygrometers Digital Thermo-Hygrometers
Rugged, handheld, portable unit is ideal for monitoring conditions for chambers, cleanrooms, food processing, labs, and all humidity-sensitive manufacturing operations.
  • Traceable® for Humidity
  • Reads °F and °C
  • Recorder Output Capability
  • L×H× D 6.4 x 26 x 2.5 cm
Thermometer can be used with chart recorder, strip recorder, or computer that accepts millivolt signals
Bright, four-digit, 1.3 cm (1/2") high LCD is readable from up to 3.1m (10') away. Permits instant readings of relative humidity and temperature; sensor updates the display 21/2 times per second. Data hold switch freezes the display to capture reading.
A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Output jack provides a 1mv signal per degree or per 1% relative humidity. Recorder output provides a hard copy record.
Ordering information: One-year warranty. Supplied with a durable carrying case and replaceable 9V alkaline battery for one year of intermittent use or 100 hours of continuous readings.