VWR® Traceable® Digital Conductivity Bench Meter
Supplier: VWR International
61161-362
61161-362EA 2767.92 USD
61161-362
VWR® Traceable® Digital Conductivity Bench Meter
Electrochemistry Meters Conductivity Meters
Designed to measure conductivity in water analysis, biotechnology, life sciences, environmental studies, and more.
  • Traceable®
  • VWR® one-year limited warranty
Can also be used to check water purity in stills and demineralizers, test laboratory glassware, measure total dissolved solids, and make up solutions.
Large, 4-digit, 12.7 mm (¹/₂") LCD shows temperature in °C and °F. Instant-response probe contains platinum electrodes and a solid-state thermometer for automatic/manual temperature compensation. All readings are referenced to the international standard of 25 °C. Computer RS232 output and recorder jack allows for continuous monitoring and a permanent record. Unit has keys to calibrate to four different calibration points. Chemical-resistant ABS housing withstands harsh environments.
Accurately provides results in conductivity (micromhos/cm and microsiemens/cm), resistivity (megohms), and dissolved solids (parts per million).
Certifications: Meets all accreditation requirements and U.S. Government specifications. Measures reagent-grade water for CAP, ASTM, NCCLS, and ACS. A serial numbered certificate is provided from an ISO 17025 calibration laboratory to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with glass probe, 115 V AC adapter, 9 V battery, receptacle to supply probe replatinizing current, and probe holder arm.