VWR® Traceable® High/Low Memory Alarm Thermometer
Supplier: VWR International
90008-24
61161-336EA 150.08 USD
61161-336
VWR® Traceable® High/Low Memory Alarm Thermometer
Thermometers Digital Thermometers
Perfect for monitoring liquids, air/gas, or semi-solids in water baths, refrigerators, freezers, incubators, and tanks.
  • Traceable®
  • Reads ambient or probe temperature
  • Minimum/maximum memory recordings
  • May be set in 1° increments
  • Displays °C and °F
Minimum/maximum memory permits monitoring overnight, weekends, or any time period.
Easy-to-view 1.3 cm (1/2") high LCD shows current, high, and low temperatures. Alarm sounds when the temperature rises above or falls below the programmable setpoints. Alarm continues to sound for up to 12 hours, even if temperature returns to non-alarm range. Sensor and 3m (10') cable perform accurately even when both are underwater. High-impact ABS plastic case is chemical-resistant.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: One-year warranty. Supplied with a flip-open stand for the lab bench, a slot for wall mounting, Velcro® fastener to attach to any surface, a wire mounting bracket for easy sensor placement, and a AA alkaline battery for continuous one-year use.