VWR® Traceable® Jumbo-Display Dial Thermometers
Supplier: VWR International
61161-310 77776-720
61161-310EA 90.06 USD
61161-310 77776-720
VWR® Traceable® Jumbo-Display Dial Thermometers
Thermometers Digital Thermometers
Thermometers fit cuvettes, test tubes, flasks, and beakers and will read temperatures in samples, reagents, water baths, foods, gases, and semi-solids.
  • Traceable®
  • Wide temperature range
  • Easy-to-read LCD
  • Read °C and °F
Readings update every second. Stainless steel probe is resistant to most laboratory chemicals. Dial has 5.4 cm (21/8") diameter and features a large, bright 1 cm (3/8") high LCD.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability toMore Product Information