VWR® Traceable® Water-Resistant Thermometer
Supplier: VWR International
46610-024 61161-283 10171-332 10171-504 10171-620
46610-024EA 105.34 USD
46610-024 61161-283 10171-332 10171-504 10171-620
VWR® Traceable® Water-Resistant Thermometer
Thermometers Digital Thermometers
Thermometers are ideal for round-the-clock monitoring of semi-solids, air/gas, or liquids in incubators, refrigerators, freezers, water baths, and outdoor tanks.
  • Traceable®
  • Waterproof
  • Shockproof
  • Stainless steel sensor
  • LCD display
Useful in wet lab/plant areas and outdoors
Thermometers may be placed anywhere. Easy-to-view 6.4mm (1/4") high LCD digits can be read at a glance. LCD operates in environments above freezing. Probe with stainless steel sensor performs accurately when under water. Records highest and lowest temperature readings over any time period. Reading updates every second.
Housing is waterproof and shockproof.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Thermometers are supplied with probe with 3m (10') cable, magnet to attach thermometer to metal, suction cups and Velcro® fastener to mount thermometer on any surface, and 1.5V silver-oxide battery for one year of continuous “always on” monitoring.