Bio-Safe NA Complete Counting Cocktail, RPI
Supplier: RPI
111198
61000-506CS 938.91 USD
61000-506
Bio-Safe NA Complete Counting Cocktail, RPI
Scintillation Cocktails
Bio-Safe NA provides the highest efficiency for counting organic samples, dry filters and samples treated with tissue solubilizers.
  • Bluish tint liquid
  • Odorless
Effective and economical cocktail for counting dry wipe tests in radiation safety procedures. Compatible with all plastic scintillation vials.
Caution: Research or further manufacturing use only, not for food or drug use.