Safety-Solve Complete Counting Cocktail, RPI
Supplier: RPI
111177
61000-504CS 995.48 USD
61000-504
Safety-Solve Complete Counting Cocktail, RPI
Scintillation Cocktails
Safety-Solve is a ready-to-use high flash point, cocktail for liquid scintillation counting.
This low-odor, high flash point, low viscosity formula does not require dilution prior to use. Contains a blend of high quantum efficiency quench resistant fluors in a non-ionic emulsifier system which will accommodate a wide range of samples.
Safety-solve complete counting cocktail is a bluish tint, liquid with slight odor.