VWR® Drying Rack, Polystyrene
Supplier: VWR International
60996-012
60996-012EA 841.82 USD
60996-012
VWR® Drying Rack, Polystyrene
Racks Draining Racks
High-impact polystyrene rack has holes to fit 72 pegs.
  • Color: Light blue
Lightweight and versatile, it accommodates cylinders, flasks, and test tubes. Pegs snap out easily. With 7.6 cm (3") draining ledge.
Ordering information: Supplied with seventy-two 10.2 cm (4") pegs, mounting kit, fixing template, and drainage tube. Additional pegs for use with test tubes are also available.