Drying Rack, Vinyl-Coated Steel Wire, Accurate Wirecraft Company
Supplier: Accurate Wirecraft
901807BK
60996-009EA 470.3 USD
60996-009
Drying Rack, Vinyl-Coated Steel Wire, Accurate Wirecraft Company
Racks Draining Racks
For draining and drying test tubes, beakers, flasks, and other laboratory glassware
Can be used flat, hung on wall, or placed upright on table or sink. Saves glassware from chipping and breaking. The vinyl coating is resistant to many chemicals and oils. Holds 90 pieces of glassware. Contains seventy-two 7.6cm (3") pins and eighteen 12.7cm (5") pins. Made of strong, extruded, steel wire, spot-welded throughout. All drainage is caught in the rustproof pan.