VWR® Signature™ PVC Tubing, Clear
Supplier: VWR International
60985-512 60985-528 60985-530 60985-532 60985-534 60985-540 60985-544 60985-584
60985-512EA 78.3 USD
60985-512 60985-528 60985-530 60985-532 60985-534 60985-540 60985-544 60985-584
VWR® Signature™ PVC Tubing, Clear
Tubing Transfer Tubing
VWR Signature™ Clear PVC Tubing is engineered for optimum performance in a wide variety of applications.
  • Lab-grade clear plastic tubing
  • Wide operating temperature range
  • Autoclavable
Designed for use in medical research, clinical, pharmaceutical, food and beverage, media manufacture and transfer, and vapor transfer applications
Durometer hardness: Shore A, 55. Operating temperature range: –32 to 71 °C (–25 to 160 °F). Tensile strength: 1650 psi. Complies with FDA, USP Class VI and USDA standards.
Can also be used with peristaltic pumps, CO₂ incubators, and condenser coolant lines. This autoclavable, lab-grade clear plastic tubing is dimensionally stable and highly resistant to hardening and discoloration. Made from the highest grade resins and special plasticizers (no fillers or extenders are added). Resists a wide range of chemicals.
Ordering information: Supplied in 50' coil lengths.