Anti-PANK1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7159A
60943-130EA 801.16 USD
60943-130
Anti-PANK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PANK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR