Anti-ACVR2B Rabbit Polyclonal Antibody

Supplier: Abgent
AP7105A
60943-064EA 808.65 USD
60943-064
Anti-ACVR2B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ACVR2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR