Anti-Creatine Kinase MB (CKM) Rabbit Polyclonal Antibody

Supplier: Abgent
AP7073A
60942-946EA 801.16 USD
60942-946
Anti-Creatine Kinase MB (CKM) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Creatine Kinase MB (CKM)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR