Anti-Aurora 3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7000G
60942-888EA 801.6 USD
60942-888
Anti-Aurora 3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Aurora 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center (100–150 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR