Anti-RCAN3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6310A
60942-668EA 801.16 USD
60942-668
Anti-RCAN3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RCAN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR