Anti-MLLT2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6189B
60942-638EA 763.01 USD
60942-638
Anti-MLLT2 Rabbit Polyclonal Antibody
Antibodies
High-quality antibodies are widely used for autophagy, apoptosis, protein partitioning, phosphorylation, cell function, and disease and gene regulation research. Antibodies are meant for research use only. They should not be used for diagnostic purposes. Antibodies are 0.1mg, unless otherwise noted. For custom antibodies, contact your VWR representative.

Type:
Antigen: MLLT2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR