Anti-RAD9 Rabbit Polyclonal Antibody

Supplier: Abgent
AP3231A
60942-610EA 801.6 USD
60942-610
Anti-RAD9 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR