Anti-Integrin beta-1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP3132A
60942-566EA 763.43 USD
60942-566
Anti-Integrin beta-1 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-Phospho-IL3R-Y593 Reacts with Human

Type: Primary
Antigen: Phospho-IL3R-Y593
Clonality: Polyclonal
Clone:
Conjugation: (none)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR