Anti-PPP4C Rabbit Polyclonal Antibody

Supplier: Abgent
AP8475A
60941-836EA 763.01 USD
60941-836
Anti-PPP4C Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PPP4C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR