Anti-PPP1CA Rabbit Polyclonal Antibody

Supplier: Abgent
AP8472A
60941-822EA 763.01 USD
60941-822
Anti-PPP1CA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PPP1CA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR