Anti-PPP2R1A Rabbit Polyclonal Antibody

Supplier: Abgent
AP8462A
60941-786EA 801.16 USD
60941-786
Anti-PPP2R1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PPP2R1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR