Anti-DUSP10 Rabbit Polyclonal Antibody

Supplier: Abgent
AP8453B
60941-758EA 763.01 USD
60941-758
Anti-DUSP10 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DUSP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR