Anti-Rat MKP3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP8445A
60941-730EA 801.6 USD
60941-730
Anti-Rat MKP3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Rat MKP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR