Anti-CDC14A Rabbit Polyclonal Antibody

Supplier: Abgent
AP8440A
60941-722EA 801.16 USD
60941-722
Anti-CDC14A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDC14A
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR