Anti-PTPN1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP8411E
60941-696EA 801.16 USD
60941-696
Anti-PTPN1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PTPN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: 1–321 aa
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR