Anti-SMG1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP8055D
60941-616EA 763.01 USD
60941-616
Anti-SMG1 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-SMG1 (N-term G14) Reacts with Human

Type: Primary
Antigen: SMG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR