Anti-SULT1C2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2602B
60941-528EA 801.16 USD
60941-528
Anti-SULT1C2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SULT1C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR