Anti-TPST1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2321A
60941-500EA 763.01 USD
60941-500
Anti-TPST1 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-TPST1 (N-term) Reacts with Human

Type: Primary
Antigen: TPST1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR