Anti-LLGL2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2199B
60941-484EA 801.16 USD
60941-484
Anti-LLGL2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LLGL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR