Anti-Reptin/TIP49B/RUVB2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1922B
60941-430EA 763.01 USD
60941-430
Anti-Reptin/TIP49B/RUVB2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Reptin/TIP49B/RUVB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR