Anti-RCC1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1919A
60941-418EA 763.01 USD
60941-418
Anti-RCC1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RCC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR